Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028244.2 | NP_082520.2 | 2 | 490 | ||
GenBank mRNA | BC016569.1 | - | 2 | 490 | 3157173 |