Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028283.2 | NP_082559.1 | 5 | 475 | ||
GenBank mRNA | AB030647.1 | - | 5 | 551 | ||
AK087466.1 | - | 5 | 516 | |||
AK148900.1 | - | 5 | 582 | |||
BC033470.1 | - | 5 | 398 | 4951775 | ||
BC042415.1 | - | 5 | 475 | 4165488 |