Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phykpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028398.2 | NP_082674.1 | 2 | 187 | ||
GenBank mRNA | AK013489.1 | - | 2 | 141 | ||
AK085984.1 | - | 2 | 138 | |||
AK133235.1 | - | 134 | ||||
AK156866.1 | - | 2 | 188 | |||
AK157759.1 | - | 2 | 137 | |||
AK160557.1 | - | 2 | 156 | |||
BC024461.1 | - | 2 | 141 | 4975449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK087703.1 | - | 134 | Chr. - - [-] on Build |