Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019746.4 | NP_062720.1 | 4, 5 | 377 | ||
GenBank mRNA | AF161074.1 | - | 4, 5 | 266 | ||
AK007297.1 | - | 4, 5 | 290 | |||
AK008632.1 | - | 4, 5 | 391 | |||
BC048476.1 | - | 4, 5 | 277 | 6773378 | ||
BC056167.1 | - | 4, 5 | 269 | 5712632 | ||
BC092092.1 | - | 4, 5 | 377 | 3492310 |