Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202469.1 | NP_001189398.1 | 3 | 280 | ||
NM_001483.2 | NP_001474.1 | 4 | 326 | |||
GenBank mRNA | AF029786.1 | - | 4 | 301 | ||
AJ001259.1 | - | 4 | 293 | |||
AK097049.1 | - | 3 | 266 | |||
AK125036.1 | - | 1 | 2582 | |||
AK315648.1 | - | 4 | 311 | |||
BC000732.2 | - | 4 | 299 | 2959662 | ||
BC001837.2 | - | 4 | 299 | 2959662 | ||
BC030821.1 | - | 4 | 308 | 4808962 | ||
BT007112.1 | - | 4 | 293 | |||
CR407612.1 | - | 4 | 293 | |||
CX166452.1 | - | 4 | 326 |