Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025346.1 | NP_079622.1 | 8, 9 | 1313 | ||
GenBank mRNA | AK005344.1 | - | 5, 6 | 692 | ||
AK075599.1 | - | 8, 9 | 1313 | |||
AK179668.1 | - | 2, 3 | 175 | |||
BC016075.1 | - | 8, 9 | 1167 | 4485143 |