Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026641.2 | NP_080917.1 | 11, 12 | 1335 | ||
GenBank mRNA | AK042231.1 | - | 11, 12 | 1260 | ||
AK129339.1 | - | 11, 12 | 1263 | |||
AK147305.1 | - | 10, 11 | 4862 | |||
BC013814.1 | - | 6, 7 | 624 | |||
BC034101.1 | - | 11, 12 | 1230 | 4501451 |