Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285826.1 | NP_001272755.1 | 5 | 948 | ||
NM_001285827.1 | NP_001272756.1 | 5 | 613 | |||
NM_027351.3 | NP_081627.1 | 5 | 427 | |||
NM_027374.3 | NP_081650.2 | 5 | 613 | |||
GenBank mRNA | AF315803.1 | - | 5 | 561 | ||
AK010207.1 | - | 5 | 431 | |||
AK011000.1 | - | 5 | 382 | |||
BC060993.1 | - | 5 | 395 | 6591278 | ||
BC061645.1 | - | 5 | 891 | 5032617 | ||
DV043111.1 | - | 5 | 649 |