Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phykpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028398.2 | NP_082674.1 | 10, 11 | 1244 | ||
GenBank mRNA | AK013489.1 | - | 10, 11 | 1198 | ||
AK133235.1 | - | 1191 | ||||
AK156866.1 | - | 10, 11 | 1589 | |||
AK160557.1 | - | 10, 11 | 1213 | |||
BC024461.1 | - | 10, 11 | 1198 | 4975449 |