Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110002H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029623.2 | NP_083899.1 | 1, 2 | 169 | ||
GenBank mRNA | AK010309.1 | - | 1, 2 | 163 | ||
AK144623.1 | - | 1, 2 | 170 | |||
AK154240.1 | - | 1, 2 | 164 | |||
AK159691.1 | - | 1, 2 | 164 | |||
BC012285.1 | - | 1, 2 | 147 | |||
BC021846.1 | - | 1, 2 | 152 | 5027035 |