Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139063.1 | NP_620702.1 | 1, 2 | 181 | ||
GenBank mRNA | AF426433.1 | - | 1, 2 | 181 | ||
AK039050.1 | - | 1, 2 | 133 | |||
AK075808.1 | - | 1, 2 | 127 | |||
AK147762.1 | - | 161 | ||||
AK170614.1 | - | 1, 2 | 131 | |||
AK171537.1 | - | 165 | ||||
BC023184.1 | - | 1, 2 | 200 | 4945742 | ||
BC024720.1 | - | 1, 2 | 138 | 5361934 |