Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205336.1 | NP_001192265.1 | 2 | 471 | ||
NM_139206.2 | NP_631945.2 | 2 | 271 | |||
GenBank mRNA | AF469621.1 | - | 1 | 221 | ||
AF469622.1 | - | 2 | 412 | |||
AK036638.1 | - | 2 | 236 | |||
AK053122.1 | - | 1 | 661 | |||
AK220486.1 | - | 2 | 264 | |||
BC068145.1 | - | 2 | 259 | 6856630 | ||
CF743699.1 | - | 2 | 296 |