Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145507.2 | NP_663482.2 | 3 | 354 | ||
NM_177445.5 | NP_803228.2 | 3 | 354 | |||
GenBank mRNA | AK077533.1 | - | 3 | 277 | ||
AK077820.1 | - | 3 | 308 | |||
AK144470.1 | - | 3 | 299 | |||
AK146350.1 | - | 3 | 298 | |||
AK151027.1 | - | 3 | 298 | |||
AK151897.1 | - | 3 | 265 | |||
AK152304.1 | - | 3 | 265 | |||
AK152562.1 | - | 3 | 265 | |||
AK153256.1 | - | 3 | 265 | |||
AK153299.1 | - | 3 | 265 | |||
AK166009.1 | - | 3 | 265 | |||
AK169247.1 | - | 3 | 265 | |||
AK169716.1 | - | 3 | 242 | |||
BB576692.2 | - | 3 | 308 | |||
BC008638.1 | - | 3 | 252 | 3586278 | ||
BY276340.1 | - | 3 | 355 | |||
BY308833.1 | - | 3 | 352 |