Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083917.1 | NP_001077386.1 | 3 | 531 | ||
NM_001286614.1 | NP_001273543.1 | 3 | 527 | |||
NM_178227.4 | NP_839941.1 | 3 | 531 | |||
GenBank mRNA | AK049286.1 | - | 3 | 529 | ||
AK049747.1 | - | 3 | 350 | |||
AK051805.1 | - | 3 | 467 | |||
AK076466.1 | - | 2 | 175 | |||
AK173115.1 | - | 3 | 408 | |||
AY049036.1 | - | 2 | 136 | |||
BC032253.1 | - | 3 | 325 | 5368626 | ||
BC053919.1 | - | 2 | 426 | 6308278 | ||
BC058083.1 | - | 3 | 470 | |||
BC058636.1 | - | 3 | 530 | 5702879 | ||
CB520894.1 | - | 3 | 531 |