Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168321.1 | NP_001161793.1 | 4 | 1071 | ||
NM_029510.3 | NP_083786.2 | 4 | 1740 | |||
NM_175044.3 | NP_778209.2 | 4 | 1740 | |||
NM_175045.3 | NP_778210.2 | 4 | 1740 | |||
NM_175046.3 | NP_778211.2 | 4 | 1740 | |||
GenBank mRNA | AK053309.1 | - | 4 | 1741 | ||
AK086385.2 | - | 4 | 1247 | |||
AK129398.1 | - | 4 | 1071 | |||
AY161170.1 | - | 4 | 1055 | |||
AY161171.1 | - | 4 | 1055 | |||
AY161172.1 | - | 4 | 1055 | |||
AY161173.1 | - | 4 | 1055 | |||
BC058656.1 | - | 4 | 1241 | 6412868 |