Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030734.2 | NP_109659.2 | 2 | 140 | ||
GenBank mRNA | AF318068.1 | - | 140 |