Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkfb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177752.1 | NP_001171223.1 | 5 | 799 | ||
NM_001177753.1 | NP_001171224.1 | 5 | 799 | |||
NM_001177754.1 | NP_001171225.1 | 5 | 799 | |||
NM_001177755.1 | NP_001171226.1 | 5 | 799 | |||
NM_001177756.1 | NP_001171227.1 | 5 | 799 | |||
NM_001177757.1 | NP_001171228.1 | 5 | 799 | |||
NM_001177758.1 | NP_001171229.1 | 5 | 436 | |||
NM_133232.3 | NP_573495.2 | 5 | 799 | |||
GenBank mRNA | AF294617.1 | - | 5 | 734 | ||
AK054285.1 | - | 5 | 784 | |||
AK077210.1 | - | 5 | 385 | |||
AK143330.1 | - | 5 | 406 | |||
AK154607.1 | - | 5 | 749 | |||
BC052400.1 | - | 5 | 776 | 5721049 | ||
BC057320.1 | - | 5 | 751 | 6826100 | ||
EU034675.1 | - | 5 | 409 | |||
EU034676.1 | - | 5 | 409 | |||
EU034677.1 | - | 5 | 409 | |||
EU034678.1 | - | 5 | 409 | |||
EU034679.1 | - | 5 | 409 |