Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133925.2 | NP_598686.2 | 8 | 974 | ||
GenBank mRNA | AK156773.1 | - | 8 | 911 | ||
AK161670.1 | - | 8 | 940 | |||
AK167050.1 | - | 8 | 976 | |||
BC018373.1 | - | 4 | 348 | 3708613 | ||
BC024602.1 | - | 8 | 855 | |||
BC027123.1 | - | 1 | 280 | 5356457 | ||
BC040811.1 | - | 8 | 919 | 4015830 |