Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276764.1 | NP_001263693.1 | 17 | 2090 | ||
NM_010081.2 | NP_034211.2 | 4 | 531 | |||
NM_133833.3 | NP_598594.2 | 14 | 1509 | |||
NM_134448.3 | NP_604443.2 | 14 | 1509 | |||
GenBank mRNA | AF396877.1 | - | 4 | 522 | ||
AF396878.1 | - | 14 | 1521 | |||
AF396879.1 | - | 14 | 1521 | |||
AK141319.1 | - | 1 | 1200 | |||
U22452.1 | - | 14 | 1509 | |||
U25158.1 | - | 17 | 2034 |