Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138750.2 | NP_620089.2 | 5, 6 | 777 | ||
NM_178047.4 | NP_835148.2 | 5, 6 | 777 | |||
GenBank mRNA | AF269062.1 | - | 5, 6 | 827 | ||
AK029851.1 | - | 5, 6 | 758 | |||
AK137749.1 | - | 5, 6 | 755 | |||
BC039212.1 | - | 5, 6 | 745 | 4188940 |