Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178061.5 | NP_835162.1 | 2 | 839 | ||
GenBank mRNA | AK039841.1 | - | 2 | 685 | ||
AK087285.1 | - | 2 | 840 | |||
AK155217.1 | - | 2 | 840 | |||
AK170973.1 | - | 2 | 833 | |||
BC020028.1 | - | 2 | 821 | 4007373 |