Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146025.2 | NP_666137.1 | 6, 7 | 1380 | ||
GenBank mRNA | AK044765.1 | - | 6, 7 | 909 | ||
AK047184.1 | - | 6, 7 | 1357 | |||
AK049753.1 | - | 6, 7 | 1383 | |||
BC034054.1 | - | 6, 7 | 904 | 4505400 | ||
BC049954.1 | - | 6, 7 | 937 | 4191611 |