Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146062.3 | NP_666174.2 | 6, 7 | 750 | ||
NM_175363.4 | NP_780572.1 | 5, 6 | 543 | |||
GenBank mRNA | AK081606.1 | - | 5, 6 | 536 | ||
AK150904.1 | - | 5, 6 | 494 | |||
AK160138.1 | - | 5, 6 | 516 | |||
BC031486.1 | - | 6, 7 | 703 | 3157250 |