Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam110a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289150.1 | NP_001276079.1 | 2 | 313 | ||
NM_001289151.1 | NP_001276080.1 | 2 | 330 | |||
NM_028666.3 | NP_082942.2 | 2 | 606 | |||
NM_146127.3 | NP_666239.1 | 2 | 461 | |||
GenBank mRNA | AJ277454.1 | - | 2 | 455 | ||
AK005776.1 | - | 2 | 330 | |||
AK017381.1 | - | 2 | 355 | |||
AK050928.1 | - | 2 | 415 | |||
AK154171.1 | - | 2 | 387 | |||
BC018162.1 | - | 2 | 298 | |||
BC025092.1 | - | 1 | 251 | 4039713 | ||
BC034240.1 | - | 2 | 371 | 4208017 | ||
BC094463.1 | - | 2 | 396 | 4224947 | ||
JF912498.1 | - | 1 | 158 |