Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168290.1 | NP_001161762.1 | 3 | 685 | ||
NM_172755.3 | NP_766343.3 | 3 | 685 | |||
GenBank mRNA | AF518875.1 | - | 3 | 678 | ||
AK042293.1 | - | 3 | 686 | |||
AK137294.1 | - | 3 | 289 | |||
BC057305.1 | - | 3 | 339 | 6822887 |