Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmpr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177992.2 | NP_818773.1 | 10 | 1034 | ||
GenBank mRNA | AK031004.1 | - | 10 | 1036 | ||
AK075772.1 | - | 10 | 992 | |||
AK179292.1 | - | 2 | 138 | |||
BC003886.1 | - | 10 | 993 | 3590411 | ||
BC024109.1 | - | 10 | 990 | 5149353 |