Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276764.1 | NP_001263693.1 | 28 | 3769 | ||
NM_010081.2 | NP_034211.2 | 15 | 2210 | |||
NM_133833.3 | NP_598594.2 | 25 | 3188 | |||
NM_134448.3 | NP_604443.2 | 25 | 3188 | |||
GenBank mRNA | AF396877.1 | - | 15 | 2201 | ||
AF396878.1 | - | 25 | 3200 | |||
AF396879.1 | - | 25 | 3200 |