Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205336.1 | NP_001192265.1 | 6 | 1196 | ||
NM_139206.2 | NP_631945.2 | 6 | 996 | |||
GenBank mRNA | AF469621.1 | - | 5 | 946 | ||
AF469622.1 | - | 6 | 1137 | |||
AK036638.1 | - | 6 | 961 | |||
AK053122.1 | - | 5 | 1386 | |||
AK220486.1 | - | 6 | 989 | |||
BC068145.1 | - | 6 | 984 | 6856630 |