Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mylk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139300.3 | NP_647461.3 | 32 | 5687 | ||
GenBank mRNA | AF314149.1 | - | 3 | 512 | ||
AK078556.1 | - | 13 | 2364 | |||
AK162008.1 | - | 14 | 2050 | |||
AY237726.1 | - | 32 | 5716 | |||
AY237727.2 | - | 17 | 3240 | |||
BC034209.1 | - | 7 | 834 | |||
BC037721.1 | - | 17 | 2250 | |||
BC038902.1 | - | 3 | 460 | |||
BC045197.1 | - | 17 | 3170 | |||
BC058610.1 | - | 32 | 5699 | 30056142 | ||
BC145279.1 | - | 32 | 5674 | 9053833 | ||
BC150749.1 | - | 32 | 5674 | 9087660 | ||
BC171954.1 | - | 32 | 5675 |