Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145360.2 | NP_663335.2 | 4, 5 | 796 | ||
GenBank mRNA | AK029302.1 | - | 4, 5 | 429 | ||
AK084208.1 | - | 4, 5 | 427 | |||
AK132073.1 | - | 4, 5 | 431 | |||
AK160832.1 | - | 4, 5 | 430 | |||
BC004801.1 | - | 3, 4 | 417 | 3589498 | ||
BC110313.1 | - | 4, 5 | 434 | 5343431 | ||
CT010310.1 | - | 3, 4 | 374 |