Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178061.5 | NP_835162.1 | 3, 4 | 1049 | ||
GenBank mRNA | AK039841.1 | - | 3, 4 | 895 | ||
AK155217.1 | - | 3, 4 | 1050 | |||
AK170973.1 | - | 3, 4 | 1043 | |||
BC020028.1 | - | 3, 4 | 1031 | 4007373 |