Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145856.2 | NP_665855.2 | 3 | 478 | ||
GenBank mRNA | AB116259.1 | - | 3 | 407 | ||
AF458064.1 | - | 2 | 383 | |||
AK157101.1 | - | 1 | 147 | |||
AY380822.1 | - | 3 | 478 | |||
BC145977.1 | - | 2 | 423 |