Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283028.1 | NP_001269957.1 | 6 | 954 | ||
NM_001283029.1 | NP_001269958.1 | 6 | 819 | |||
NM_001283030.1 | NP_001269959.1 | 5 | 678 | |||
NM_011462.3 | NP_035592.1 | 4 | 583 | |||
NM_146043.4 | NP_666155.1 | 6 | 879 | |||
GenBank mRNA | AK088127.1 | - | 6 | 820 | ||
AK134259.1 | - | 6 | 842 | |||
AK135847.1 | - | 4 | 582 | |||
AK135933.1 | - | 4 | 581 | |||
AK136233.1 | - | 4 | 581 | |||
AK136251.1 | - | 4 | 582 | |||
AK139899.1 | - | 4 | 580 | |||
AK162171.1 | - | 4 | 580 | |||
AK162195.1 | - | 4 | 580 | |||
AK162258.1 | - | 4 | 577 | |||
AK163263.1 | - | 4 | 579 | |||
AK163293.1 | - | 4 | 580 | |||
AK163301.1 | - | 4 | 584 | |||
AK163308.1 | - | 4 | 582 | |||
AK163382.1 | - | 4 | 580 | |||
AK166528.1 | - | 4 | 583 | |||
AK166580.1 | - | 4 | 542 | |||
AK166677.1 | - | 4 | 579 | |||
AK168544.1 | - | 6 | 880 | |||
BC016517.1 | - | 6 | 865 | 2654233 | ||
U48972.1 | - | 4 | 577 |