Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083114.1 | NP_001076583.1 | 8 | 922 | ||
NM_001285863.1 | NP_001272792.1 | 8 | 1051 | |||
NM_001285864.1 | NP_001272793.1 | 7 | 830 | |||
NM_146062.3 | NP_666174.2 | 10 | 1186 | |||
NM_175363.4 | NP_780572.1 | 9 | 979 | |||
GenBank mRNA | AK081606.1 | - | 9 | 972 | ||
AK150904.1 | - | 9 | 930 | |||
AK158488.1 | - | 8 | 1052 | |||
AK160138.1 | - | 9 | 952 | |||
AK168443.1 | - | 8 | 924 | |||
BC031486.1 | - | 10 | 1139 | 3157250 | ||
BC150854.2 | - | 7 | 826 | 9087765 |