Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284189.1 | NP_001271118.1 | 5 | 1211 | ||
NM_011982.3 | NP_036112.1 | 5 | 1211 | |||
NM_147176.3 | NP_671705.2 | 5 | 1211 | |||
NM_152134.3 | NP_687036.1 | 5 | 730 | |||
GenBank mRNA | AB019478.1 | - | 5 | 645 | ||
AB019479.1 | - | 5 | 907 | |||
AB089435.1 | - | 5 | 522 | |||
AF093257.1 | - | 5 | 1213 | |||
AF093258.1 | - | 5 | 560 | |||
AK137151.1 | - | 1950 | ||||
AY137385.1 | - | 5 | 730 | |||
BC064041.1 | - | 5 | 1031 | 1398649 |