Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136236.1 | NP_001129708.1 | 4 | 707 | ||
NM_153090.2 | NP_694730.2 | 5 | 767 | |||
NM_178165.4 | NP_835459.1 | 4 | 707 | |||
GenBank mRNA | AF329485.1 | - | 4 | 634 | ||
AK038494.1 | - | 4 | 647 | |||
AK156528.1 | - | 4 | 647 | |||
AK156985.1 | - | 4 | 647 | |||
AY158088.1 | - | 4 | 627 | |||
AY158089.1 | - | 4 | 627 | |||
AY506554.1 | - | 4 | 640 | |||
AY506555.1 | - | 4 | 640 | |||
BC055830.1 | - | 4 | 634 | 3599941 |