Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025778.1 | NP_001020949.1 | 3 | 441 | ||
NM_001308420.1 | NP_001295349.1 | 3 | 279 | |||
NM_016440.3 | NP_057524.3 | 3 | 441 | |||
XM_005258971.3 | XP_005259028.1 | 3 | 768 | |||
XM_005258972.3 | XP_005259029.1 | 3 | 768 | |||
XM_006723237.3 | XP_006723300.1 | 3 | 582 | |||
XM_006723238.3 | XP_006723301.1 | 3 | 382 | |||
XM_011527023.2 | XP_011525325.1 | 3 | 768 | |||
GenBank mRNA | AB031052.1 | - | 3 | 213 | ||
AF514788.1 | - | 3 | 273 | |||
AK225154.1 | - | 3 | 186 | |||
AK292918.1 | - | 3 | 184 | |||
AK294802.1 | - | 3 | 183 | |||
AK303010.1 | - | 3 | 216 | |||
AK303529.1 | - | 3 | 184 | |||
AK311220.1 | - | 3 | 183 | |||
BC023556.2 | - | 3 | 220 | 4298578 | ||
BC095449.1 | - | 3 | 193 | 30721912 | ||
BG421578.1 | - | 3 | 362 | |||
BP335032.1 | - | 3 | 279 |