Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173182.2 | NP_775274.2 | 23 | 2947 | ||
GenBank mRNA | AB098596.1 | - | 23 | 2885 | ||
AK136013.1 | - | 2 | 145 | |||
AK147346.1 | - | 24 | 2957 | |||
AK220297.1 | - | 23 | 2947 | |||
AY234860.1 | - | 5 | 689 | |||
BC067389.1 | - | 23 | 2953 | 30431862 | ||
DQ192037.1 | - | 22 | 2853 |