Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175353.2 | NP_780562.2 | 2 | 268 | ||
GenBank mRNA | AK082881.1 | - | 2 | 247 | ||
AK151524.1 | - | 2 | 269 | |||
AK151838.1 | - | 2 | 269 | |||
BC026859.1 | - | 2 | 227 | 2631201 | ||
BY100185.1 | - | 2 | 269 |