Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hopx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159900.1 | NP_001153372.1 | 3 | 325 | ||
NM_001159901.1 | NP_001153373.1 | 2 | 453 | |||
NM_175606.3 | NP_783199.1 | 3 | 485 | |||
GenBank mRNA | AF492703.1 | - | 3 | 485 | ||
AF492704.1 | - | 3 | 313 | |||
AF534182.1 | - | 3 | 530 | |||
AF536202.1 | - | 2 | 222 | |||
AI604115.1 | - | 2 | 439 | |||
AK003784.1 | - | 3 | 314 | |||
AK008297.1 | - | 3 | 292 | |||
AK008439.1 | - | 3 | 292 | |||
AK009007.1 | - | 3 | 242 | |||
BC024546.1 | - | 3 | 309 | 4989797 |