Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018755.2 | NP_061225.2 | 4 | 809 | ||
NM_176073.4 | NP_788262.1 | 3 | 719 | |||
GenBank mRNA | AF009513.1 | - | 3 | 600 | ||
AF107835.1 | - | 3 | 687 | |||
AK032972.1 | - | 3 | 584 | |||
AK075686.1 | - | 3 | 650 | |||
AK135096.1 | - | 3 | 706 | |||
AK137164.1 | - | 3 | 593 | |||
AK152145.1 | - | 4 | 687 | |||
AK154361.1 | - | 3 | 329 | |||
BC010977.1 | - | 3 | 704 | |||
BC017373.1 | - | 3 | 722 | |||
BC037067.1 | - | 3 | 566 | 4989536 |