Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177041.3 | NP_796015.2 | 2, 3 | 311 | ||
GenBank mRNA | AK044501.1 | - | 320 | |||
AK161929.1 | - | 2, 3 | 355 | |||
AK211873.1 | - | 1, 2 | 178 | |||
BC025817.1 | - | 2, 3 | 264 | 5099127 | ||
BC046769.1 | - | 2, 3 | 311 | 6809196 |