Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 5 | 697 | ||
NR_026691.1 | 6 | 827 | ||||
GenBank mRNA | AB209626.1 | - | 4 | 2198 | ||
AK291645.1 | - | 5 | 697 | |||
AK301260.1 | - | 4 | 515 | |||
AK301484.1 | - | 6 | 827 | |||
AK301492.1 | - | 5 | 967 | |||
AK313049.1 | - | 5 | 691 | |||
AK316146.1 | - | 5 | 749 | |||
AK316342.1 | - | 6 | 901 | |||
BC028082.1 | - | 5 | 674 | 5216986 | ||
U00672.1 | - | 5 | 681 |