Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 2 | 228 | ||
NM_177345.4 | NP_796319.1 | 2 | 554 | |||
GenBank mRNA | AK052045.1 | - | 2 | 222 | ||
AK132263.1 | - | 2 | 218 | |||
AK148428.1 | - | 2 | 543 | |||
BC043296.1 | - | 2 | 453 | |||
BC090644.1 | - | 1 | 128 | 30629263 | ||
BC096618.1 | - | 2 | 527 | 6392317 |