Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177460.4 | NP_803411.3 | 3 | 483 | ||
GenBank mRNA | AK033791.1 | - | 433 | |||
AK133475.1 | - | 3 | 484 | |||
AK166002.1 | - | 2 | 247 | |||
AK169866.1 | - | 3 | 398 | |||
BC014731.1 | - | 2 | 513 | |||
BC055447.1 | - | 3 | 452 | 3594406 |