Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D19Bwg1357e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050829.1 | - | 3 | 160 | ||
AK088081.1 | - | 3 | 156 | |||
AK141893.1 | - | 3 | 158 | |||
BC016186.1 | - | 3 | 173 | |||
BC041111.1 | - | 2 | 138 | |||
BC052686.1 | - | 2 | 112 | |||
BC145533.1 | - | 3 | 157 | |||
BY251590.1 | - | 3 | 248 | |||
BY289042.1 | - | 3 | 232 |