Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018755.2 | NP_061225.2 | 5 | 959 | ||
NM_176073.4 | NP_788262.1 | 4 | 869 | |||
GenBank mRNA | AF009513.1 | - | 4 | 750 | ||
AF107835.1 | - | 4 | 837 | |||
AK032972.1 | - | 4 | 734 | |||
AK075686.1 | - | 4 | 800 | |||
AK135096.1 | - | 4 | 856 | |||
AK137164.1 | - | 4 | 743 | |||
AK152139.1 | - | 5 | 837 | |||
AK152145.1 | - | 5 | 837 | |||
AK154361.1 | - | 4 | 479 | |||
BC010977.1 | - | 4 | 854 | |||
BC017373.1 | - | 4 | 872 | |||
BC037067.1 | - | 4 | 716 | 4989536 |