Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapkap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290625.1 | NP_001277554.1 | 4 | 509 | ||
NM_001290626.1 | NP_001277555.1 | 3 | 507 | |||
NM_177345.4 | NP_796319.1 | 4 | 835 | |||
GenBank mRNA | AK042154.1 | - | 3 | 497 | ||
AK052045.1 | - | 4 | 503 | |||
AK132263.1 | - | 4 | 499 | |||
AK148428.1 | - | 4 | 824 | |||
AK178100.1 | - | 2 | 83 | |||
BC043296.1 | - | 4 | 734 | |||
BC090644.1 | - | 3 | 409 | 30629263 | ||
BC096618.1 | - | 4 | 808 | 6392317 |