Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286809.1 | NP_001273738.1 | 2, 3 | 451 | ||
NM_009792.3 | NP_033922.1 | 3, 4 | 484 | |||
NM_177407.4 | NP_803126.1 | 14, 15 | 1172 | |||
GenBank mRNA | AK083245.1 | - | 14, 15 | 1173 | ||
AK122410.2 | - | 14, 15 | 1168 | |||
BB659027.1 | - | 2, 3 | 407 | |||
BC031745.1 | - | 14, 15 | 1153 | 4506005 | ||
X14836.1 | - | 14, 15 | 1156 | |||
X87142.1 | - | 3, 4 | 484 |