Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177789.4 | NP_808457.1 | 2 | 401 | ||
GenBank mRNA | AK041014.1 | - | 2 | 402 | ||
BC025105.1 | - | 2 | 377 | 4162875 |